Furfurylmercaptan
Lieferant: Molekula
Synonyme:
2-Furanmethanthiol, 2-Furylmethanthiol
15974779-100G
MOLE15974779-100GEA
0
EUR
MOLE15974779-100G
Furfurylmercaptan
Furfurylmercaptan
Formel:
C₅H₆OS Molecular Weight: 114,17 g/mol Siedepunkt: 155 °C (1013 hPa) Dichte: 1,1339 g/cm³ (20 °C) Flammpunkt: 51 °C |
MDL:
MFCD00003254 CAS-Nummer: 98-02-2 EINECS: 202-628-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...