2'-Deoxyinosin
Lieferant: Molekula
Synonyme:
9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol
15986561-1G
MOLE15986561-1GEA
0
EUR
MOLE15986561-1G
2'-Deoxyinosin
2'-Deoxyinosin
Formel:
C₁₀H₁₂N₄O₄ Molecular Weight: 252,23 g/mol |
MDL:
MFCD00005762 CAS-Nummer: 890-38-0 EINECS: 212-964-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...