Triethylammoniumchlorid

Lieferant: Molekula
Achtung

Synonyme: Triethylamin Hydrochlorid

16002121-1KG
MOLE16002121-1KGEA 0 EUR
MOLE16002121-1KG
Triethylammoniumchlorid
Triethylammonium chloride
Formel: C₆H₁₆ClN
Molecular Weight: 137,65 g/mol
Dichte: 0,7637 g/cm³ (20 °C)
MDL: MFCD00012500
CAS-Nummer: 554-68-7
EINECS: 209-067-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR