(+)-Di-p-toluoyl-D-weinsäure

Lieferant: Molekula

Synonyme: Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid

16022090-25G
MOLE16022090-25GEA 0 EUR
MOLE16022090-25G
(+)-Di-p-toluoyl-D-weinsäure
(+)-Di-p-toluoyl-D-weinsäure
Formel: C₂₀H₁₈O₈
Molecular Weight: 386,36 g/mol
MDL: MFCD00008552
CAS-Nummer: 32634-68-7
EINECS: 251-132-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR