1,3-Dichlor-5,5-dimethylhydantoin
Lieferant: Molekula
Synonyme:
1,3-Dichloro-5,5-dimethylhydantoine, 1,3-Dichloro-5,5-dimethylimidazolidine-2,4-dione
16229036-500G
MOLE16229036-500GEA
0
EUR
MOLE16229036-500G
1,3-Dichlor-5,5-dimethylhydantoin
1,3-Dichlor-5,5-dimethylhydantoin
Formel:
C₅H₆Cl₂N₂O₂ Molecular Weight: 197,02 g/mol |
MDL:
MFCD00003190 CAS-Nummer: 118-52-5 EINECS: 204-258-7 UN: 1479 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...