2,2-Dimethyl-1,3-dioxan-4,6-dion (Isopropylidenmalonat, Meldrumsäure)
Lieferant: Molekula
Synonyme:
2,2-Dimethyl-1,3-dioxane-4,6-dione
16238403-100G
MOLE16238403-100GEA
0
EUR
MOLE16238403-100G
2,2-Dimethyl-1,3-dioxan-4,6-dion (Isopropylidenmalonat, Meldrumsäure)
2,2-Dimethyl-1,3-dioxan-4,6-dion (Isopropylidenmalonat, Meldrumsäure)
Formel:
C₆H₈O₄ Molecular Weight: 144,13 g/mol |
MDL:
MFCD00006638 CAS-Nummer: 2033-24-1 EINECS: 217-992-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...