1,4-Phenyldiboronsäure

Lieferant: Molekula

16250319-1G
MOLE16250319-1GEA 0 EUR
MOLE16250319-1G
1,4-Phenyldiboronsäure
1,4-Phenyldiboronsäure
Formel: C₆H₈B₂O₄
MDL: MFCD00236018
CAS-Nummer: 4612-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR