Dexamethason
Lieferant: Molekula
Synonyme:
9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum
16320764-1G
MOLE16320764-1GEA
106
EUR
MOLE16320764-1G
Dexamethason
Dexamethason
Formel:
C₂₂H₂₉FO₅ Molecular Weight: 392,47 g/mol Schmelzpunkt: 268…271 °C |
MDL:
MFCD00064136 CAS-Nummer: 50-02-2 EINECS: 200-003-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...