Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Lieferant: Molekula
16368689-25G
MOLE16368689-25GEA
0
EUR
MOLE16368689-25G
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Formel:
C₁₀H₁₄N₅Na₂O₁₃P₃ |
MDL:
MFCD00080339 CAS-Nummer: 987-65-5 EINECS: 213-579-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...