Cyclobutanon
Lieferant: Molekula
16515758-5G
MOLE16515758-5GEA
356
EUR
MOLE16515758-5G
Cyclobutanon
Cyclobutanone
Formel:
C₄H₆O Molecular Weight: 70,09 g/mol Siedepunkt: 99 °C (1013 hPa) Dichte: 0,93 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001332 CAS-Nummer: 1191-95-3 EINECS: 214-745-6 UN: 1224 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...