5,5-Dimethyl-2,4-imidazolidindion

Lieferant: Molekula

Synonyme: 5,5-Dimethylimidazolidine-2,4-dione

16523938-1KG
MOLE16523938-1KGEA 176 EUR
MOLE16523938-1KG
5,5-Dimethyl-2,4-imidazolidindion
5,5-Dimethylhydantoin
Formel: C₅H₈N₂O₂
Molecular Weight: 128,13 g/mol
MDL: MFCD00005266
CAS-Nummer: 77-71-4
EINECS: 201-051-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR