(Methyl)triphenylphosphoniumbromid

Lieferant: Molekula

16542796-100G
MOLE16542796-100GEA 0 EUR
MOLE16542796-100G
(Methyl)triphenylphosphoniumbromid
(Methyl)triphenylphosphoniumbromid
Formel: C₁₉H₁₈BrP
Molecular Weight: 357.23 g/mol
Schmelzpunkt: 232
Flammpunkt: >240 °C
MDL: MFCD00011804
CAS-Nummer: 1779-49-3
EINECS: 217-218-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR