TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)

Lieferant: Molekula
Gefahr

Synonyme: TCEP HCl

16589050-1G
MOLE16589050-1GEA 71.6 EUR
MOLE16589050-1G
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)
Formel: C₉H₁₆ClO₆P
Molecular Weight: 286,65 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00145469
CAS-Nummer: 51805-45-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR