DL-α-Phenylglycin
Lieferant: Molekula
Synonyme:
Dl-A-Aminophenylacetic Acid, H-DL-Phg-OH, DL-2-Phenylglycine
16677282-500G
MOLE16677282-500GEA
201
EUR
MOLE16677282-500G
DL-α-Phenylglycin
DL-α-Phenylglycin
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol |
MDL:
MFCD00064402 CAS-Nummer: 2835-06-5 EINECS: 220-608-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...