2-Fluorbenzolsulfonylchlorid
Lieferant: Molekula
Synonyme:
2-Fluorbenzol-1-sulfonylchlorid, 2-Fluorbenzolsulfochlorid, 2-Fluorbenzolsulfonsäurechlorid
16742290-5G
MOLE16742290-5GEA
0
EUR
MOLE16742290-5G
2-Fluorbenzolsulfonylchlorid
2-Fluorbenzolsulfonylchlorid
Formel:
C₆H₄ClFO₂S Molecular Weight: 194.61 g/mol Siedepunkt: 242 Dichte: 1.47 g/cm³ (20 °C) |
MDL:
MFCD00042285 CAS-Nummer: 2905-21-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...