EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)
Lieferant: Molekula
Synonyme:
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate
16764018-100G
MOLE16764018-100GEA
157
EUR
MOLE16764018-100G
EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)
EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)
Formel:
C₁₄H₁₇NO₃ Molecular Weight: 247.29 g/mol |
MDL:
MFCD00006703 CAS-Nummer: 16357-59-8 EINECS: 240-418-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...