4,4'-Diaminobenzanilid

Lieferant: Molekula

Synonyme: 4-Aminobenzoesäure(4-aminophenyl)amid

16774468-100G
MOLE16774468-100GEA 0 EUR
MOLE16774468-100G
4,4'-Diaminobenzanilid
4,4'-Diaminobenzanilid
Formel: C₁₃H₁₃N₃O
Molecular Weight: 227.27 g/mol
Schmelzpunkt: 214…215 °C
MDL: MFCD00025361
CAS-Nummer: 785-30-8
EINECS: 212-321-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR