Dibenzo-18-krone-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecan, Zinn(II)-Ionophor I)

Lieferant: Molekula

Synonyme: Dibenzo-18-crown-6

16800887-25G
MOLE16800887-25GEA 0 EUR
MOLE16800887-25G
Dibenzo-18-krone-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecan, Zinn(II)-Ionophor I)
Dibenzo-18-krone-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecan, Zinn(II)-Ionophor I)
Formel: C₂₀H₂₄O₆
Molecular Weight: 360.41 g/mol
MDL: MFCD00005098
CAS-Nummer: 14187-32-7
EINECS: 238-041-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR