Bestatin Hydrochlorid ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucin Hydrochlorid)

Lieferant: Molekula

Synonyme: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl

16832467-5MG
MOLE16832467-5MGEA 0 EUR
MOLE16832467-5MG
Bestatin Hydrochlorid ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucin Hydrochlorid)
Bestatin Hydrochlorid ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucin Hydrochlorid)
Formel: C₁₆H₂₅ClN₂O₄
Molecular Weight: 308,4 · 36,5 g/mol
MDL: MFCD00058004
CAS-Nummer: 65391-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR