AMPSO Natriumsalz

Lieferant: Molekula

16851613-100G
MOLE16851613-100GEA 0 EUR
MOLE16851613-100G
AMPSO Natriumsalz
AMPSO Natriumsalz
Formel: C₇H₁₆NNaO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00067500
CAS-Nummer: 102029-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR