4-(Benzyloxy)anilin Hydrochlorid
Lieferant: Molekula
Synonyme:
4-Benzyloxyanilinium Chloride, 4-Aminophenyl benzyl ether hydrochloride, p-Benzyloxyaniline hydrochloride
16990076-100G
MOLE16990076-100GEA
0
EUR
MOLE16990076-100G
4-(Benzyloxy)anilin Hydrochlorid
4-(Benzyloxy)anilin Hydrochlorid
Formel:
C₁₃H₁₄ClNO Molecular Weight: 235,711 g/mol |
MDL:
MFCD00012995 CAS-Nummer: 51388-20-6 EINECS: 257-170-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...