Indol-5-carbonitril

Lieferant: Molekula

Synonyme: 1H-Indol-5-carbonitril

17057160-5G
MOLE17057160-5GEA 256 EUR
MOLE17057160-5G
Indol-5-carbonitril
Indol-5-carbonitril
Formel: C₉H₆N₂
Molecular Weight: 142,16 g/mol
Schmelzpunkt: 106…108 °C
MDL: MFCD00005669
CAS-Nummer: 15861-24-2
EINECS: 239-986-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR