ω-Brom-4-fluoracetophenon
Lieferant: Molekula
Synonyme:
2-Brom-1-(4-fluorphenyl)ethan-1-on, Brommethyl-p-fluorphenylketon, 2-Brom-4'-fluoracetophenon, 4-Fluorphenacylbromid, α-Brom-4-fluoracetophenon
17125731-25G
MOLE17125731-25GEA
0
EUR
MOLE17125731-25G
ω-Brom-4-fluoracetophenon
ω-Brom-4-fluoracetophenon
Formel:
C₈H₆BrFO Molecular Weight: 217,04 g/mol Siedepunkt: 82 °C (0,4 mmHg) Schmelzpunkt: 49 °C |
MDL:
MFCD00040830 CAS-Nummer: 403-29-2 EINECS: 206-955-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...