Aminoacetaldehyddiethylacetal
Lieferant: Molekula
Synonyme:
2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal
17211839-100G
MOLE17211839-100GEA
0
EUR
MOLE17211839-100G
Aminoacetaldehyddiethylacetal
Aminoacetaldehyddiethylacetal
Formel:
C₆H₁₅NO₂ Molecular Weight: 133,19 g/mol |
MDL:
MFCD00008136 CAS-Nummer: 645-36-3 EINECS: 211-439-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...