2,2,2-Trifluorethylammoniumchlorid
Lieferant: Molekula
Synonyme:
1-Amino-2,2,2-trifluorethan Hydrochlorid, 2,2,2-Trifluorethylamin Hydrochlorid
17542068-25G
MOLE17542068-25GEA
0
EUR
MOLE17542068-25G
2,2,2-Trifluorethylammoniumchlorid
2,2,2-Trifluorethylammoniumchlorid
Formel:
C₂H₅ClF₃N Molecular Weight: 135,52 g/mol Schmelzpunkt: 222…226 °C |
MDL:
MFCD00012875 CAS-Nummer: 373-88-6 EINECS: 206-771-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...