1-(3-Methoxyphenyl)piperazin Dihydrochlorid

Lieferant: Molekula

17665229-25G
MOLE17665229-25GEA 0 EUR
MOLE17665229-25G
1-(3-Methoxyphenyl)piperazin Dihydrochlorid
1-(3-Methoxyphenyl)piperazin Dihydrochlorid
Formel: C₁₁H₁₈Cl₂N₂O
Molecular Weight: 265,18 g/mol
MDL: MFCD00035285
CAS-Nummer: 6968-76-9
EINECS: 230-189-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR