6-Methoxyindol
Lieferant: Molekula
|
Achtung
|
Synonyme:
6-Methoxy-1H-indol, Indol-6-ylmethylether
17682700-1G
MOLE17682700-1GEA
0
EUR
MOLE17682700-1G
6-Methoxyindol
6-Methoxyindol
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00022780 CAS-Nummer: 3189-13-7 EINECS: 221-689-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...