2-Amino-5-mercapto-1,3,4-thiadiazol
Lieferant: Molekula
Synonyme:
5-Amino-1,3,4-thiadiazole-2-thiol
17804191-100G
MOLE17804191-100GEA
0
EUR
MOLE17804191-100G
2-Amino-5-mercapto-1,3,4-thiadiazol
2-Amino-5-mercapto-1,3,4-thiadiazole
Formel:
C₂H₃N₃S₂ Molecular Weight: 133,2 g/mol |
MDL:
MFCD00003108 CAS-Nummer: 2349-67-9 EINECS: 219-078-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...