N,N,N',N'-Tetramethyl-p-phenylendiamin
Lieferant: Molekula
|
Achtung
|
18154918-25G
MOLE18154918-25GEA
0
EUR
MOLE18154918-25G
N,N,N',N'-Tetramethyl-p-phenylendiamin
N,N,N',N'-Tetramethyl-p-phenylendiamin
Formel:
C₁₀H₁₆N₂ Siedepunkt: 260 °C (1013 hPa) Schmelzpunkt: 48…50 °C Dichte: 1,08 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008309 CAS-Nummer: 100-22-1 EINECS: 202-831-6 UN: 1673 ADR: 6.1,III Merck Index: 12,09367 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...