(4-(Brommethyl)phenyl)essigsäure

Lieferant: Molekula

Synonyme: 4-(Bromomethyl)phenylacetic acid

18181226-1G
MOLE18181226-1GEA 0 EUR
MOLE18181226-1G
(4-(Brommethyl)phenyl)essigsäure
(4-(Brommethyl)phenyl)essigsäure
Formel: (C₆H₄(CH₂Br))CH₂COOH
Molecular Weight: 229,07 g/mol
MDL: MFCD00010632
CAS-Nummer: 13737-36-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR