7-Hydroxyindol

Lieferant: Molekula

18308008-1G
MOLE18308008-1GEA 0 EUR
MOLE18308008-1G
7-Hydroxyindol
7-Hydroxyindol
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
MDL: MFCD00152102
CAS-Nummer: 2380-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR