3,5-Dichlorsalicylsäure

Lieferant: Molekula

Synonyme: 3,5-Dichlor-2-hydroxybenzoesäure

18351091-100G
MOLE18351091-100GEA 0 EUR
MOLE18351091-100G
3,5-Dichlorsalicylsäure
3,5-Dichlorsalicylsäure
Formel: C₇H₄Cl₂O₃
Molecular Weight: 207,01 g/mol
Siedepunkt: 319 °C (1013 hPa)
MDL: MFCD00002442
CAS-Nummer: 320-72-9
EINECS: 206-281-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR