7-Methylxanthin

Lieferant: Molekula

Synonyme: 7-Methyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dihydro-7-methyl-1H-purin-2,6-dion, 7-Methyl-1H,3H-purin-2,6-dion, 2,6-Dihydroxy-7-methylpurin, 7-Methyl-3,7-dihydro-2H-purin-2,6-dion, 7-Methyl-2,6(1H,3H)-purindion

18393522-100MG
MOLE18393522-100MGEA 139 EUR
MOLE18393522-100MG
7-Methylxanthin
7-Methylxanthin
Formel: C₆H₆N₄O₂
Molecular Weight: 166.14 g/mol
MDL: MFCD00037979
CAS-Nummer: 552-62-5
EINECS: 209-019-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR