6-Fluorindol
Lieferant: Molekula
|
Achtung
|
Synonyme:
6-Fluoro-1H-indole
18394914-1G
MOLE18394914-1GEA
71.6
EUR
MOLE18394914-1G
6-Fluorindol
6-Fluorindol
Formel:
C₈H₆FN Molecular Weight: 135.14 g/mol Schmelzpunkt: 72…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056933 CAS-Nummer: 399-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...