4-Chlor-2-fluoranilin
Lieferant: Molekula
18590478-25G
MOLE18590478-25GEA
0
EUR
MOLE18590478-25G
4-Chlor-2-fluoranilin
4-Chlor-2-fluoranilin
Formel:
C₆H₅ClFN Molecular Weight: 145,56 g/mol Siedepunkt: 104 °C (1013 hPa) Dichte: 1,299 g/cm³ (20 °C) |
MDL:
MFCD00010625 CAS-Nummer: 57946-56-2 EINECS: 261-034-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...