2-Methoxybenzylalkohol

Lieferant: Molekula

Synonyme: (2-Methoxyphenyl)methanol, 2-(Hydroxymethyl)anisol

18967483-100G
MOLE18967483-100GEA 0 EUR
MOLE18967483-100G
2-Methoxybenzylalkohol
2-Methoxybenzylalkohol
Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Siedepunkt: 250 °C (1013 hPa)
Dichte: 1,12 g/cm³ (20 °C)
MDL: MFCD00004611
CAS-Nummer: 612-16-8
EINECS: 210-296-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR