Histamin Dihydrochlorid

Lieferant: Molekula

Synonyme: Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride

19012772-25G
MOLE19012772-25GEA 131 EUR
MOLE19012772-25G
Histamin Dihydrochlorid
Histamin Dihydrochlorid
Formel: C₅H₁₁Cl₂N₃
Molecular Weight: 184,07 g/mol
MDL: MFCD00012703
CAS-Nummer: 56-92-8
EINECS: 200-298-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR