Triethylphosphonoformiat

Lieferant: Molekula

19063440-500G
MOLE19063440-500GEA 0 EUR
MOLE19063440-500G
Triethylphosphonoformiat
Triethylphosphonoformiat
Formel: C₇H₁₅O₅P
MDL: MFCD00009108
CAS-Nummer: 1474-78-8
EINECS: 216-016-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR