(Methyl)triphenylphosphoniumiodid

Lieferant: Molekula

Synonyme: Methyltriphenylphosphonium iodide

19063878-100G
MOLE19063878-100GEA 196 EUR
MOLE19063878-100G
(Methyl)triphenylphosphoniumiodid
(Methyl)triphenylphosphoniumiodid
Formel: C₁₉H₁₈IP
Molecular Weight: 404,23 g/mol
Schmelzpunkt: 193…194 °C
MDL: MFCD00066175
CAS-Nummer: 2065-66-9
EINECS: 218-178-5
UN: 3464
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR