5-Chlor-2-fluorbenzoesäure

Lieferant: Molekula
Achtung

19115143-5G
MOLE19115143-5GEA 0 EUR
MOLE19115143-5G
5-Chlor-2-fluorbenzoesäure
5-Chlor-2-fluorbenzoesäure
Formel: C₇H₄ClFO₂
Molecular Weight: 174,56 g/mol
MDL: MFCD00665762
CAS-Nummer: 394-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR