Nystatin Dihydrat

Lieferant: Molekula

Synonyme: Fungicidin dihydrate, Mycostatin dihydrate

19268873-5G
MOLE19268873-5GEA 122 EUR
MOLE19268873-5G
Nystatin Dihydrat
Nystatin Dihydrat
Formel: C₄₇H₇₅NO₁₇·2H₂O
Molecular Weight: 926,09 g/mol (anhy)
MDL: MFCD00036240
CAS-Nummer: 1400-61-9
EINECS: 215-749-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR