Aminoguanidiniumhydrogencarbonat

Lieferant: Molekula

Synonyme: Aminoguanidine hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine hydrogencarbonate, Guanylhydrazine hydrogencarbonate

19361491-500G
MOLE19361491-500GEA 116 EUR
MOLE19361491-500G
Aminoguanidiniumhydrogencarbonat
Aminoguanidinium hydrogen carbonate
Formel: C₂H₈N₄O₃
Molecular Weight: 136,11 g/mol
MDL: MFCD00012949
CAS-Nummer: 2582-30-1
EINECS: 219-956-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR