DL-Isoleucin

Lieferant: Molekula

Synonyme: H-DL-Ile-OH

19387777-5G
MOLE19387777-5GEA 60.8 EUR
MOLE19387777-5G
DL-Isoleucin
DL-Isoleucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131,17 g/mol
Schmelzpunkt: 275 °C
Dichte: 1,255 g/cm³ (–123.15 °C)
MDL: MFCD00004268
CAS-Nummer: 443-79-8
EINECS: 207-139-8
Merck Index: 13,05198

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR