4-Benzyloxyindol

Lieferant: Molekula

19412115-1G
MOLE19412115-1GEA 0 EUR
MOLE19412115-1G
4-Benzyloxyindol
4-Benzyloxyindol
Formel: C₁₅H₁₃NO
Molecular Weight: 223,27 g/mol
MDL: MFCD00047200
CAS-Nummer: 20289-26-3
EINECS: 243-690-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR