(1-Pentyl)triphenylphosphoniumbromid

Lieferant: Molekula
Achtung

Synonyme: Pentyltriphenylphosphonium bromide

19517443-25G
MOLE19517443-25GEA 0 EUR
MOLE19517443-25G
(1-Pentyl)triphenylphosphoniumbromid
(1-Pentyl)triphenylphosphoniumbromid
Formel: H₃C(CH₂)₄P(C₆H₅)₃Br
Molecular Weight: 413,34 g/mol
Schmelzpunkt: 168 °C
MDL: MFCD00031630
CAS-Nummer: 21406-61-1
EINECS: 244-374-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bf324e98e27624d1eab0e430f6eee7a96fa960&vid=e2bf324e98e27624d1eab0e430f6eee7a96fa960&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR