5-Chlor-2-indolcarbonsäure
Lieferant: Molekula
Synonyme:
5-Chloro-1H-indole-2-carboxylic acid
19584892-5G
MOLE19584892-5GEA
0
EUR
MOLE19584892-5G
5-Chlor-2-indolcarbonsäure
5-Chloroindole-2-carboxylic acid
Formel:
C₉H₆ClNO₂ Schmelzpunkt: 291…292 °C |
MDL:
MFCD00005613 CAS-Nummer: 10517-21-2 EINECS: 234-050-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...