2,4,5,6-Tetraaminopyrimidinsulfat

Lieferant: Molekula

19915065-25G
MOLE19915065-25GEA 0 EUR
MOLE19915065-25G
2,4,5,6-Tetraaminopyrimidinsulfat
2,4,5,6-Tetraaminopyrimidinsulfat
Formel: C₄H₈N₆·H₂SO₄
Molecular Weight: 238,23 g/mol
MDL: MFCD00012786
CAS-Nummer: 5392-28-9
EINECS: 226-393-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR