Triphenylphosphoniumbromid

Lieferant: Molekula

Synonyme: Triphenylphosphine hydrobromide

20107888-25G
MOLE20107888-25GEA 0 EUR
MOLE20107888-25G
Triphenylphosphoniumbromid
Triphenylphosphonium bromide
Formel: C₁₈H₁₆BrP
Molecular Weight: 343,2 g/mol
MDL: MFCD00035107
CAS-Nummer: 6399-81-1
EINECS: 229-012-6
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR