2-Amino-4,6-dichlorpyrimidin
Lieferant: Molekula
Synonyme:
4,6-Dichloropyrimidin-2-ylamine
20239572-5G
MOLE20239572-5GEA
0
EUR
MOLE20239572-5G
2-Amino-4,6-dichlorpyrimidin
2-Amino-4,6-dichloropyrimidine
Formel:
C₄H₃Cl₂N₃ Molecular Weight: 163,99 g/mol Schmelzpunkt: 216…220 °C |
MDL:
MFCD00006090 CAS-Nummer: 56-05-3 EINECS: 200-253-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...