N-Acetyl-β-D-glucosamin

Lieferant: Molekula

Synonyme: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

20293490-100G
MOLE20293490-100GEA 0 EUR
MOLE20293490-100G
N-Acetyl-β-D-glucosamin
N-Acetyl-β-D-glucosamin
Formel: C₈H₁₅NO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00136044
CAS-Nummer: 7512-17-6
EINECS: 231-368-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR